missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coenzyme Q10, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
Description
Coenzyme Q10 is an essential cofactor of the electron transport chain and a potent antioxidant agent.
Specifications
Specifications
| Chemical Name or Material | Coenzyme Q10 |
| CAS | 303-98-0 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C59H90O4 |
| Quantity | 1 g |
| Synonym | CoQ10 Ubiquinone-10 |
| Solubility Information | DMF : 40 mg/mL (46.33 mM; Need ultrasonic) ∣Ethanol : 2.5 mg/mL (2.90 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.16 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=C(C)C(C(OC)=C1OC)=O |
| Molecular Weight (g/mol) | 863.34 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction