missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt, TRC

Product Code. 30134603
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134603 1 g 1g
30007865 100 mg 100mg
30011180 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134603 Supplier Toronto Research Chemicals Supplier No. TRCC6364001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18439-24-2
Molecular Formula C21 H33 N7 O16 P3 S . 3 Li
Molecular Weight (g/mol) 785.33
Synonym CoA Trilithium Salt, Trilithium Coenzyme A;
IUPAC Name trilithium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)[O-])n2cnc3c(N)ncnc23)C(O)C(=O)NCCC(=O)NCCS

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H36N7O16P3S.3Li/c1-21(2,16(31)19(32)24-4-3-12(29)23-5-6-48)8-41-47(38,39)44-46(36,37)40-7-11-15(43-45(33,34)35)14(30)20(42-11)28-10-27-13-17(22)25-9-26-18(13)28;;;/h9-11,14-16,20,30-31,48H,3-8H2,1-2H3,(H,23,29)(H,24,32)(H,36,37)(H,38,39)(H2,22,25,26)(H2,33,34,35);;;/q;3*+1/p-3/t11-,14-,15-,16?,20-;;;/m1.../s1
Formula Weight 785.1397
Percent Purity >95%
Chemical Name or Material Coenzyme A Trilithium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.