missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate, 96%, Fisher BioReagents

Product Code. 10226394
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10226394 1 g Amber Glass 1g
10174814 50 mg Amber Glass 50mg
10032804 250 mg Amber Glass 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10226394 Supplier Fisher BioReagents Supplier No. BP25101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 18439-24-2
Molecular Formula C21H33Li3N7O16P3S
Molecular Weight (g/mol) 785.329
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
Synonym coenzyme a, trilithium salt
PubChem CID 53442190
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O

Specifications

Color White
Assay Percent Range ≥96 %
Packaging Amber Glass
Quantity 1 g
Percent Purity 96% min.
Physical Form Powder or Solid
Chemical Name or Material Coenzyme A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.