missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A, MedChemExpress

Product Code. 16478147
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16478147

Brand: Medchem Express HY128851

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Coenzyme A, a ubiquitous essential cofactor, is an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism. Coenzyme A plays a central role in the metabolism of carboxylic acids, including short- and long-chain fatty acids.

Coenzyme A, a ubiquitous essential cofactor, is an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism. Coenzyme A plays a central role in the metabolism of carboxylic acids, including short- and long-chain fatty acids.

Specifications

Chemical Name or Material Coenzyme A
CAS 85-61-0
Color White
Purity Grade Notes Research
Molecular Formula C21H36N7O16P3S
Quantity 5 mg
Solubility Information H2O : 125 mg/mL (162.86 mM; Need ultrasonic) ∣DMSO : 16.25 mg/mL (21.17 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](O[C@H](COP(OP(OCC(C)([C@@H](O)C(NCCC(NCCS)=O)=O)C)(O)=O)(O)=O)[C@H]1OP(O)(O)=O)N2C3=NC=NC(N)=C3N=C2
Molecular Weight (g/mol) 767.53
Formula Weight 767.53
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.