missing translation for 'onlineSavingsMsg'
Learn More

Melford Coenzyme A

Product Code. 30018201
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
This item is not returnable. View return policy

Product Code. 30018201

Brand: Melford C708000.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extracted from yeast.

Extracted from yeast. Coenzyme A is a coenzyme, notable for its role in the synthesis and oxidation of fatty acids, and the oxidation of pyuvate in the citric acid cycle.

Specifications

CAS 85-61-0
Quantity 100 mg
Molecular Formula C21H36N7O16P3S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.