Learn More
DiscoverX Coelenterazine h
Description
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 10mg of dry powder.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Biochemical Reagent |
| Target Entry | Calcium |