Learn More
DiscoverX Coelenterazine h
Product Code. 17372339
Encompass
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
17372339 1 mg 1mg
1 options
Product Code. 17372339 Supplier DiscoverX Supplier No. 900084
Coelenterazine h is a modified coelenterazine substrate for aequorin-based calcium assays. This product contains 1mg of dry powder.
This product is an optimized form of coelenterazine substrate that is significantly more sensitive to calcium, intended for use in aequorin-based luminescent calcium assays. This product is shipped as a dry powder. This product contains 1mg of dry powder.
Specifications
Quantity 1 mg
Product Type Biochemical Reagent
Target Entry Calcium