missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Coelenterazine

Product Code. 16437618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437618 10 mg 10mg
16433616 25 mg 25mg
16430088 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16437618 Supplier MedChemExpress Supplier No. HY18743
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Coelenterazine is a luminescent enzyme substrate for apoaequorin and Renilla luciferase.

Renilla luciferase and substrate coelenterazine has been used as the bioluminescence donor in bioluminescence resonance energy transfer (BRET) to detect protein-protein interactions. Coelenterazine is a superoxide anion-sensitive chemiluminescent probe and it can also be used in chemiluminescent detection of peroxynitrite.

Specifications

Quantity 10 mg
Storage Requirements -20°C, protect from light, stored under nitrogen ∣ The compound is unstable in solutions, freshly prepared is recommended.
Solubility DMSO : 50 mg/mL (118.07 mM; Need ultrasonic; DMSO can inactivate Coelenterazine's activity) ∣ Ethanol : 2 mg/mL (4.72 mM; Need ultrasonic; DMSO can inactivate Coelenterazine's activity)
Shipping Condition Room temperature
CAS 55779-48-1
Molecular Weight (g/mol) 423.46 g/mol
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.