Learn More
Brady™ Codesoft Lite USB Hardware
Beskrivning
Codesoft Lite is for use with Brady™ Label Printers. Compatible with BBP™11 Label Printer, BBP™12 Industrial Label Printer, BBP™72 Double-sided Sleeve Printer, BradyPrinter PR Plus Series Label Printer, IP™ Series Printer, PAM3000 Print and Apply Machine, PAM3600 Print and Apply Machine, THT 76 mm Core Printers, and TLS-PC LINK™ Thermal Labeling System.
This enterprise-level label design software allows users to design and print custom labels for product identification and tracking. Codesoft software is the professional solution for the most complex barcode labeling projects. The software provides a range of barcode label design features for any project, from easy label designs to general barcoding. This software will only work when the hardware key is connected.
Requires:
- 32 or 64 bit Windows operating systems: 7, XP, Server 2003 or 2008 or Vista
- Free USB port
- 1 GB of free RAM
- Hard drive with at least 1 GB free disk space (depending on the installation options)
- CPU: 2 Ghz minimum
- Video card: DirectX 9 minimum
Specifikationer
Specifikationer
| For Use With (Equipment) | Brady Label Printers |
| Languages | Arabic, Czech, Danish, Dutch, English, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Spanish, Swedish, Turkish |
| Software Type | USB Key |
| Operating System | Windows™ 7 |
| For Use With (Application) | Asset Tracking, Barcode Labeling, Inventory and Inspection Labeling |
| Warranty | Service Contract: 3 Year |
| Alternate Part Number | CS LT USB 3 SMA |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.