Learn More
Gilson™ Code 24 Tube Rack
Brand: Gilson™ 150427
Additional Details : Weight : 0.45000kg
Description
- These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments.
- The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates.
Specifications
Tube Rack | |
Polypropylene | |
1 Pack |
Tubes (28 mm) | |
14 | |
28 mm |