missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Coconut oil, pure, refined

Product Code. 10262422 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10262422 500 g Glass bottle 500g
10169841 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262422 Supplier Thermo Scientific Chemicals Supplier No. 365475000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8001-31-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-31-8
MDL Number MFCD00130817

Specifications

Acidity 0.5mg KOH/g max.
Melting Point 23.0°C to 26.0°C
Color White
Density 0.9030g/mL
Flash Point >200°C
Packaging Glass bottle
Quantity 500 g
Specific Gravity 0.903
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide
Grade Pure
Physical Form Unctious Mass
Chemical Name or Material Coconut oil, Refined
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.