missing translation for 'onlineSavingsMsg'
Learn More

Coconut Oil, MP Biomedicals

Product Code. 11485502 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 lb.
Unit Size:
1lb

Product Code. 11485502

Brand: MP Biomedicals 0290140301

Please to purchase this item. Need a web account? Register with us today!

Major components: Tripalmitin, Tristearin, Trimyristin, Trilaurin

TRUSTED_SUSTAINABILITY

Specifications

CAS 8001-31-8
Quantity 1 lb.
Chemical Name or Material Coconut Oil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.