missing translation for 'onlineSavingsMsg'
Learn More

Coconut Oil, MP Biomedicals™

Product Code. 11495502 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 lb.
25 lb.
Unit Size:
25lb
5lb

Product Code. 11495502

Brand: MP Biomedicals 0290140305

Please to purchase this item. Need a web account? Register with us today!

Coconut Oil, White Waxy Solid to Off-White Hazy Viscous liquid

  • It is considered an edible oil and has several purposes in food, industry, and medicine.
  • Naturally occurring lipid which contains high saturated fat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-31-8
MDL Number MFCD00130817
Synonym Coconut fat, Coconut oil from Cocos nucifera, Copra oil

Specifications

Melting Point 80.6°F (literature)
Color White
Flash Point >235.4°F/ >113°C (Lit.)
Quantity 5 lb.
Physical Form Waxy Solid to Hazy Viscous Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.