missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COBRA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527602
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (86%), Xenopus laevis (85%), Pig (97%), Chicken (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).
Specifications
| COBRA1 | |
| Polyclonal | |
| Unconjugated | |
| NELFB | |
| A730008L03Rik; AB041607; AI663983; Cobra1; cofactor of BRCA1; DKFZp586B0519; KIAA1182; MNCb-5210; Negative elongation factor B; negative elongation factor complex member B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELFB; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1); RGD1307832; RP13-122B23.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25920, 58202 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| Q8C4Y3, Q8WX92 | |
| NELFB | |
| Recombinant protein fragment corresponding to a region within amino acids 1 and 206 of Human COBRA1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction