Learn More
Ekastu Safety™ Cobra Foldy FFP1 Filtering Facepiece
Product Code. 10526920
Change view
Click to view available options
Product Type:
General Purpose Particulate Respirator
Unit Size:
Pack of 20
Product Code. Product Type Unit Size
10526920 General Purpose Particulate Respirator Pack of 20
1 options
Product Code. 10526920 Supplier Ekastu Safety™ Supplier No. 419210
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protects against fine inert particles up to 4.5 times the NPF value. Ekastu Safety™ Cobra Foldy FFP1 Filtering Facepiece has a five-year shelf life in original packaging.
  • Construction of mask in 3D panel design
  • Foam lined comfort nose zone
  • Flexible nose clip
  • With comfortable, cycled harness and unique comfort-harness clip
  • Metal-free band holder-strap
  • Specially designed for wearer of EKASTU spectacles and goggles
  • Space saving due to fold ability
  • Individually packed to avoid contamination during storage
  • Protection against fine inert particles up to 4.5 times the NPF value
  • Protection class DIN EN 149:2001 FFP1 NR
  • In original packaging: 5 years shelf life
  • 20 pieces per pack
    Specifications
    Certifications/Compliance DIN EN 149:2001 FFP1 NR
    Product Type General Purpose Particulate Respirator
    For Use With (Application) Specially designed for wearers of EKASTU spectacles and goggles
    Quantity 20/Pk.
    Style Cycled Harness and Unique Comfort-harness Clip