missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COBLL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560868
This item is not returnable.
View return policy
Description
Immunogen sequence: NNSAHNEQNS QIPTPTDGPS FTVMRQSSLT FQSSDPEQMR QSLLTAIRSG EAAAKLKRVT IPSNTISVNG RSRLSHSMSP D Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 73%.
The function remains known.
Specifications
| COBLL1 | |
| Polyclonal | |
| Unconjugated | |
| COBLL1 | |
| 1810047P18Rik; Cobll1; COBL-like 1; Coblr1; cobl-related 1; cobl-related protein 1; Cordon-bleu protein-like 1; Cordon-bleu protein-like 1-like protein; cordon-bleu WH2 repeat protein like 1; cordon-bleu WH2 repeat protein-like 1; D430044D16Rik; KIAA0977 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22837 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53SF7 | |
| COBLL1 | |
| Recombinant protein corresponding to Human COBLL1. Recombinant protein control fragment (Product #RP-99630). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction