missing translation for 'onlineSavingsMsg'
Learn More

Cobimetinib (R-enantiomer), MedChemExpress

Product Code. 16468247
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468247 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468247 Supplier Medchem Express Supplier No. HY13079
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cobimetinib R-enantiomer is the less active R-enantiomer of Cobimetinib. Cobimetinib is a potent and selective MEK inhibitor.

Cobimetinib R-enantiomer is the less active R-enantiomer of Cobimetinib. Cobimetinib is a potent and selective MEK inhibitor.

Specifications

Chemical Name or Material Cobimetinib (R-enantiomer)
CAS 934660-94-3
Color Pink
Purity Grade Notes Research
Molecular Formula C21H21F3IN3O2
Quantity 5 mg
Synonym GDC-0973 R-enantiomer XL-518 R-enantiomer
Solubility Information DMSO : ≥ 100 mg/mL (188.21 mM)
SMILES FC1=C(F)C=CC(C(N2CC([C@H]3CCCCN3)(O)C2)=O)=C1NC4=CC=C(I)C=C4F
Molecular Weight (g/mol) 531.31
Formula Weight 531.31
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.