missing translation for 'onlineSavingsMsg'
Learn More

Cobalt wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 11321498
Click to view available options
Quantity:
2 m
50 cm
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 cm
Percent Purity 99.995%
Grade Puratronic™
Odor Odorless
Weight ≈1.75g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Cobalt wire, 0.5mm (0.02 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'