missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Sulphate Heptahydrate, TRC

Product Code. 30351733
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
30351733 5 g -
30355075 10 g 10g
30354614 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 30351733

Brand: Toronto Research Chemicals TRCC7250435G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
IUPAC Name cobalt(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Co+2].[O-]S(=O)(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/Co.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2
Formula Weight 280.9589
Chemical Name or Material Cobalt Sulphate Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.