missing translation for 'onlineSavingsMsg'
Learn More

Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3

Product Code. 10246490 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10246490

Brand: Thermo Scientific Acros 195925000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cobalt standard solution
Name Note 1 mg/ml Co in 2% HNO3
Packaging Plastic bottle
CAS 10141-05-6, 7732-18-5
Physical Form Solution
Molecular Formula CoN2O6
Concentration 990 to 1010 μg/mL of Co(2+) in 2 to 5% HNO3
MDL Number MFCD00010935
Solubility Miscible with water
InChI Key UFMZWBIQTDUYBN-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2]
IUPAC Name cobalt(2+);dinitrate
Molecular Weight (g/mol) 182.941
PubChem CID 25000
Quantity 500 mL
ChEBI CHEBI:86209
Formula Weight 58.93
Grade AAS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cobalt atomic absorption standard solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.