missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Standard: Co @ 5000 μg/g in Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
50 g
200 g
400 g
800 g
Unit Size:
200g
400g
50g
800g
Description
Cobalt Standard: Co @ 5000 μg/g in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Cobalt |
| CAS | 7440-48-4 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Cobalt |
| Shipping Condition | Room Temperature |
| Molecular Formula | Co |
| Certifications/Compliance | ISO 17034 |
| Synonym | Co |
| SMILES | [Co] |
| IUPAC Name | Cobalt |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction