missing translation for 'onlineSavingsMsg'
Learn More

Cobalt slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11354458
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]
This item is not returnable. View return policy
7440-48-4
Co
58.93
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
104730
CHEBI:27638
cobalt
[Co]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Percent Purity 99.95%
Odor Odorless
Weight ≈0.45g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Cobalt slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'