missing translation for 'onlineSavingsMsg'
Learn More

Cobalt phthalocyanine, MedChemExpress

Product Code. 16471749
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471749 Supplier Medchem Express Supplier No. HY18761
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cobalt phthalocyanine is a catalyst of redox reaction, catalyzes aerobic regenerations of aldehydes and ketones from aldoximes and ketoximes has been developed.

Cobalt phthalocyanine is a catalyst of redox reaction, catalyzes aerobic regenerations of aldehydes and ketones from aldoximes and ketoximes has been developed.

Specifications

Chemical Name or Material Cobalt phthalocyanine
CAS 3317-67-7
Color Pale Purple
Purity Grade Notes Research
Molecular Formula C32H16CoN8
Quantity 1 g
Synonym Cobalt(II) phthalocyanine Cobaltous phthalocyanine Phthalocyanine cobalt complex
Solubility Information DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES [N-]12[Co+2]34[N-]5C6=NC(C7=CC=CC=C87)=[N]3C8=NC1=C(C=CC=C9)C9=C2N=C%10[N]4=C(C%11=CC=CC=C%10%11)N=C5C%12=CC=CC=C6%12
Molecular Weight (g/mol) 571.46
Formula Weight 571.46
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.