missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(IV) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11381529
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381529 25 g 25g
11391529 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381529 Supplier Thermo Scientific Alfa Aesar Supplier No. 014005.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cobalt(IV) sulfide is a promising battery electrode material exhibiting good electrochemical properties for lithium sulfur batteries, thermal batteries and lithium ion secondary battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12013-10-4
Molecular Formula CoS2
Molecular Weight (g/mol) 123.05
MDL Number MFCD00064735
InChI Key XUKVMZJGMBEQDE-UHFFFAOYSA-N
Synonym cobalt 3+ ;trisulfide, cobalt sulfide,
PubChem CID 25147475
SMILES S=[Co]=S

Specifications

Color Black
Physical Form Powder
Quantity 25 g
UN Number UN3077
IUPAC Name disulfanylidenecobalt
Formula Weight 123.06
Percent Purity 99.5%
Odor Odorless
Assay (metals basis excluding Ni), Ni <0.2%
Chemical Name or Material Cobalt(IV) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.