missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(III) 2,4-pentanedionate

Product Code. 11357377
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11357377

Brand: Thermo Scientific Alfa Aesar 012536.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21679-46-9 | C15H21CoO6 | 356.26 g/mol

Cobalt(III) 2,4-pentanedionate is used as a polymerization catalyst, vulcanizing agent, pigment in resins and glass. It is also used for the preparation of monoporphyrinates. Further, it is used in kinetic studies of iron-sulfur proteins. In addition to this, it reacts with trimethylaluminum to prepare Me(3Al/Co(acac)3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21679-46-9
Molecular Formula C15H21CoO6
Molecular Weight (g/mol) 356.26
MDL Number MFCD00013488
InChI Key VACIXPOMJJIDBM-UHFFFAOYSA-N
Synonym Cobalt(III) acetylacetonate
IUPAC Name 2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5⁶.5⁶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide
SMILES CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4

Specifications

Melting Point 211°C (decomposition)
Odor Light
Quantity 100 g
Sensitivity Hygroscopic; Moisture sensitive
Solubility Information Soluble in methanol and halogenated solvents. Slightly soluble in acetone,aromatic solvents and aliphatic solvents. Insoluble in water.
Formula Weight 356.26
Physical Form Powder
Chemical Name or Material Cobalt(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.