missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) tungsten oxide, 99%

Product Code. 11349377
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349377 10 g 10g
11369377 50 g 50g
11359377 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349377 Supplier Thermo Scientific Alfa Aesar Supplier No. 040223.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

pigment, drier for enamels, inks, and paints, electronic devices | CAS: 10101-58-3 | CoO4W | 306.77 g/mol

It is used as a pigment, drier for enamels, inks, and paints, electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-58-3
Molecular Formula CoO4W
Molecular Weight (g/mol) 306.77
MDL Number MFCD00016034
InChI Key BGLJOHZOBUGBBY-UHFFFAOYSA-N
Synonym cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten
PubChem CID 11738337
IUPAC Name cobalt(2+);dioxido(dioxo)tungsten
SMILES [O--].[O--].[O--].[O--].[Co++].[W]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water; Soluble in hot concentrated acids
Formula Weight 306.78
Percent Purity 99%
Density 8.42 g/mL
Chemical Name or Material Cobalt(II) tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.