missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals
51.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
It is used as a catalysts for the industrial process called hydrodesulfurization. It is used to purify the cobalt from its ore & in qualitative inorganic analysis. It is used as a electrode material for lithium sulfur batteries, thermal batteries & lithium ion secondary batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >1116°C |
Color | Red |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.5% (Metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 91 |
Odor | Odorless |
Appearance | Red powder |
Chemical Name or Material | Cobalt(II) sulfide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'