missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate hydrate, Reagent, Co 20.8% min

Product Code. 11331109
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331109 250 g 250g
11321109 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331109 Supplier Thermo Scientific Alfa Aesar Supplier No. 011340.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier | CAS: 60459-08-7 | CoO4S | 154.99 g/mol

Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60459-08-7
Molecular Formula CoO4S
Molecular Weight (g/mol) 154.99
MDL Number MFCD00149657
InChI Key KTVIXTQDYHMGHF-UHFFFAOYSA-L
Synonym cobalt ii sulfate hydrate, cobalt sulfate monohydrate, unii-5zqa69klc0, 5zqa69klc0, cobalt mesosulfate coh2so5, cobalt sulfate coso4 monohydrate, cobalt ii sulfatehydrate, cobalt sulphate monohydrate, co.so4.h2o, cobalt 2+ hydrate sulfate
PubChem CID 16210984
IUPAC Name cobalt(2+);sulfate;hydrate
SMILES [Co++].[O-]S([O-])(=O)=O

Specifications

Color Red to Orange
Physical Form Crystalline
Quantity 250 g
Linear Formula CoSO4·xH2O
UN Number UN3077
Solubility Information Soluble in water; Slightly soluble in methanol and ethanol
Formula Weight 154.99 (Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) Co 20.8% min.
Chemical Name or Material Cobalt(II) sulfate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.