missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) sulfate hydrate, Reagent, Co 20.8% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red to Orange |
Physical Form | Crystalline |
Quantity | 1 kg |
Linear Formula | CoSO4·xH2O |
UN Number | UN3077 |
Solubility Information | Soluble in water; Slightly soluble in methanol and ethanol |
Formula Weight | 154.99 (Anhydrous) |
Grade | Reagent |
Concentration or Composition (by Analyte or Components) | Co 20.8% min. |
Chemical Name or Material | Cobalt(II) sulfate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'