missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfamate hydrate

Product Code. 11394948
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394948 2 kg 2kg
11384948 100 g 100g
11304958 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394948 Supplier Thermo Scientific Alfa Aesar Supplier No. 043906.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14017-41-5 | CoH4N2O6S2 | 251.09 g/mol

Cobalt(II) sulfamate hydrate is used in photonic device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14017-41-5
Molecular Formula CoH4N2O6S2
Molecular Weight (g/mol) 251.09
MDL Number MFCD00049877
InChI Key WLQXLCXXAPYDIU-UHFFFAOYSA-L
Synonym cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek
PubChem CID 26398
SMILES [Co++].NS([O-])(=O)=O.NS([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 2 kg
Solubility Information Soluble in water.
IUPAC Name λ²-cobalt(2+) disulfamate
Formula Weight 251.11 (Anhydrous)
Odor Odorless
Chemical Name or Material Cobalt (II) sulfamate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.