missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) stearate, Co 9-10%, Thermo Scientific™

Product Code. 11379518
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
20 kg
Unit Size:
1kg
20kg
250g
50g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379518 20 kg 20kg
11399518 50 g 50g
11389518 250 g 250g
11309528 1 kg 1kg
11319528 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11379518 Supplier Thermo Scientific Alfa Aesar Supplier No. 043352.B8
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-88-6
Molecular Formula C36H70CoO4
Molecular Weight (g/mol) 625.89
MDL Number MFCD00054343
InChI Key AMFIJXSMYBKJQV-UHFFFAOYSA-L
Synonym cobalt ii stearate, cobaltous stearate, cobalt stearate, stearic acid cobalt ii salt, octadecanoic acid, cobalt salt, cobalt distearate, cobalt 2+ dioctadecanoate, stearic acid cobalt salt, cobalt 2+ ; octadecanoate
PubChem CID 6451168
IUPAC Name cobalt(2+);octadecanoate
SMILES [Co++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

Color Purple
Density 1.71 g/mL
Linear Formula (C18H35O2)2Co
Quantity 20 kg
Solubility Information Insoluble in water.
Formula Weight 625.89
Concentration or Composition (by Analyte or Components) Co 9 to 10%
Physical Form Pellets
Chemical Name or Material Cobalt (II) stearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.