missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) selenide, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11389377
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389377 250 g 250g
11379377 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389377 Supplier Thermo Scientific Alfa Aesar Supplier No. 040224.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1307-99-9
Molecular Formula CoSe
Molecular Weight (g/mol) 137.904
MDL Number MFCD00016020
InChI Key QVYIMIJFGKEJDW-UHFFFAOYSA-N
Synonym cobalt selenide, cobalt ii selenide, cobalt selenide, cose, cobalt ii selenide trace metals basis 50g
PubChem CID 102108
IUPAC Name selanylidenecobalt
SMILES [Co]=[Se]

Specifications

Physical Form Powder
Quantity 250 g
UN Number UN3283
Solubility Information Soluble in HNO3,aqua regia
Formula Weight 137.89
Percent Purity ≥99%
Odor Weak
Density 7.650 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) selenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.