missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phosphate octahydrate

Product Code. 11392598
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11392598

Brand: Thermo Scientific Alfa Aesar 023111.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-50-5 | Co3H16O16P2 | 510.859 g/mol

It is used as a pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-50-5
Molecular Formula Co3H16O16P2
Molecular Weight (g/mol) 510.859
MDL Number MFCD00016028
InChI Key QOQSIXJUYVUEMP-UHFFFAOYSA-H
Synonym cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate
PubChem CID 202398
IUPAC Name cobalt(2+);diphosphate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Slightly soluble in water. Soluble in mineral acids.
Formula Weight 510.87 (366.74 Anhydrous)
Odor Odorless
Density 2.769 g/mL
Chemical Name or Material Cobalt(II) phosphate octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.