missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phosphate, anhydrous, 98%

Product Code. 11479567
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479567 25 g 25g
11489567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479567 Supplier Thermo Scientific Alfa Aesar Supplier No. B22066.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13455-36-2 | Co3O8P2 | 366.739 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-36-2
Molecular Formula Co3O8P2
Molecular Weight (g/mol) 366.739
MDL Number MFCD00016028
InChI Key ZBDSFTZNNQNSQM-UHFFFAOYSA-H
Synonym cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
PubChem CID 61615
IUPAC Name cobalt(2+);diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]

Specifications

Quantity 25 g
Formula Weight 366.74
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Cobalt(II) phosphate, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.