missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phosphate, anhydrous, 98%

Product Code. 11489567
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11489567

Brand: Thermo Scientific Alfa Aesar B22066.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13455-36-2 | Co3O8P2 | 366.739 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-36-2
Molecular Formula Co3O8P2
Molecular Weight (g/mol) 366.739
MDL Number MFCD00016028
InChI Key ZBDSFTZNNQNSQM-UHFFFAOYSA-H
Synonym cobalt phosphate, cobaltous phosphate, cobalt ii phosphate, tricobalt bis orthophosphate, unii-0b5m38t47h, cobalt ii phosphate, anhydrous, phosphoric acid, cobalt 2+ salt 2:3, 3co.2po4, cobalt ii phosphate octahydrate, phosphoric acid,cobalt 2+ salt 2:3
PubChem CID 61615
IUPAC Name cobalt(2+);diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]

Specifications

Quantity 100 g
Formula Weight 366.74
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Cobalt(II) phosphate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.