missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxide, 99.995% (metals basis)

Product Code. 11478500
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478500 25 g 25g
11488500 5 g 5g
11468500 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478500 Supplier Thermo Scientific Alfa Aesar Supplier No. 044354.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1307-96-6 | CoO | 74.93 g/mol

Cobalt(II) oxide is widely used in the ceramic industry. It is an additive to create blue-colored glazes and enamels. It finds application in the manufacture of magnetic and electronic component materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1307-96-6
Molecular Formula CoO
Molecular Weight (g/mol) 74.93
MDL Number MFCD00016031
InChI Key IVMYJDGYRUAWML-UHFFFAOYSA-N
Synonym cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13
PubChem CID 14786
IUPAC Name oxocobalt
SMILES O=[Co]

Specifications

Melting Point 1935°C
Color Brown-Green
Physical Form Powder
Quantity 25 g
UN Number UN2811
Merck Index 14,2446
Solubility Information Insoluble in water.
Formula Weight 74.93
Percent Purity 99.995%
Concentration or Composition (by Analyte or Components) CoO, may contain up to 10% Co3O4
Sensitivity Air Sensitive
Density 6.45 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt (II) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.