missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) oxide, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Pigments, glass coloring, semiconductors. Cobalt(II) oxide is widely used in ceramic industries. It is an additive to create blue-colored glazes and enamels. It finds application in the manufacture of magnetic and electronic component materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1935°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 95% |
Solubility Information | Soluble in acids or alkalis. Insoluble in water. |
Formula Weight | 74.93 |
Sensitivity | Air Sensitive |
Density | 6.45 |
Chemical Name or Material | Cobalt(II) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'