missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate dihydrate, Reagent Grade

Product Code. 11365029
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365029 100 g 100g
11375029 5 g 5g
11395029 25 g 25g
11385029 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11365029 Supplier Thermo Scientific Alfa Aesar Supplier No. 087758.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stabilizer for HCN, temperature indicator | CAS: 5965-38-8 | C2H4CoO6 | 182.981 g/mol

Cobalt(II) oxalate dihydrate is used as a stabilizer for hydrogen cyanide and as a temperature indicator. It is used in the preparation of cobalt catalysts, and cobalt metal powder for powder-metallurgical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 5965-38-8
Molecular Formula C2H4CoO6
Molecular Weight (g/mol) 182.98
MDL Number MFCD03414148
InChI Key MWHSMSAKVHVSAS-UHFFFAOYSA-L
Synonym cobaltousoxalate,cobaltoxalate
PubChem CID 131881712
IUPAC Name 1,3-dioxa-2-cobaltacyclopentane-4,5-dione dihydrate
SMILES O.O.O=C1O[Co]OC1=O

Tekniske data

Melting Point 229°C to 231°C (decomposition)
Density 3.021 g/mL
Odor Odorless
Quantity 100 g
UN Number UN3288
Merck Index 14,2445
Solubility Information Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids.
Formula Weight 182.98 (146.95 Anhydrous)
Grade Reagent
Physical Form Powder
Chemical Name or Material Cobalt (II) oxalate dihydrate
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.