missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate dihydrate, Puratronic™, 99.995% (metals basis)

Product Code. 11372108
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372108 50 g 50g
11382108 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372108 Supplier Thermo Scientific Alfa Aesar Supplier No. 013012.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5965-38-8 | C2H4CoO6 | 182.981 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-38-8
Molecular Formula C2H4CoO6
Molecular Weight (g/mol) 182.98
MDL Number MFCD03414148
InChI Key MWHSMSAKVHVSAS-UHFFFAOYSA-L
Synonym cobaltousoxalate,cobaltoxalate
PubChem CID 131881712
IUPAC Name 1,3-dioxa-2-cobaltacyclopentane-4,5-dione dihydrate
SMILES O.O.O=C1O[Co]OC1=O

Specifications

Melting Point 229°C to 231°C (decomposition)
Color Pink
Physical Form Powder
Quantity 50 g
Linear Formula CoC2O4·2H2O
UN Number UN3288
Merck Index 14,2445
Formula Weight 182.98 (146.95 Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Odor Odorless
Density 3.021 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) oxalate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.