missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate, anhydrous, 98%

Product Code. 11456106
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11456106

Brand: Thermo Scientific Alfa Aesar B22956.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 814-89-1 | C2CoO4 | 146.951 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-89-1
Molecular Formula C2CoO4
Molecular Weight (g/mol) 146.951
MDL Number MFCD00050793
InChI Key MULYSYXKGICWJF-UHFFFAOYSA-L
Synonym cobalt oxalate, cobalt oxalate 1:1, cobalt oxalate coc2o4, cobalt; oxalic acid, oxalic acid cobalt 2+ salt 1:1, ethanedioic acid, cobalt 2+ salt 1:1, oxalic acid cobalt, cobalt ii oxalate,anhydrous, oxalic acid, cobalt 2+ salt 1:1
PubChem CID 69946
IUPAC Name cobalt(2+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Co+2]

Specifications

Melting Point >300°C
Quantity 10 g
Merck Index 14,2445
Formula Weight 146.95
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Cobalt(II) oxalate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.