missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent

Product Code. 10573134 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10573134

Brand: Thermo Scientific Acros 423580025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55.0°C
Color Brown-Pink to Red
Physical Form Adhering Crystalline Powder or Crystals
Quantity 2.5 kg
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula Co(NO3)2·6H2O
Merck Index 15, 2428
Solubility Information Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents
Formula Weight 291.02
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 1.88
Density 1.8800g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.