missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) iodide, ultra dry, 99.999% (metals basis)

Product Code. 15447455
Change view
Click to view available options
Quantity:
1 g
Packaging:
Ampouled under argon
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15447455 1 g Ampouled under argon 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447455 Supplier Thermo Scientific Alfa Aesar Supplier No. 047185.03
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15238-00-3 | CoI2

Cobalt(II) iodide is used as a moisture and humidity indicator. It is also a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15238-00-3
Melting Point 515°C (vac.)
Boiling Point 570°C (vac.)
Packaging Ampouled under argon
Molecular Formula CoI2
MDL Number MFCD00016030
Quantity 1 g
Sensitivity Hygroscopic; Light sensitive
Synonym cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis
Solubility Information Soluble in water.
Formula Weight 312.74
Percent Purity 99.999%
Physical Form Powder
Chemical Name or Material Cobalt(II) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.