missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II,III) oxide

Product Code. 11443157
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443157 250 g 250g
11433157 50 g 50g
11423157 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443157 Supplier Thermo Scientific Alfa Aesar Supplier No. A16121.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1308-06-1 | Co3O4 | 240.80 g/mol

Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Quantity 250 g
UN Number UN3077
Merck Index 14,2429
Solubility Information Insoluble in water.
Formula Weight 240.8
Density 6.11
Chemical Name or Material Cobalt(II,III) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.