missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II,III) oxide, 99.7% (metals basis)

Product Code. 11329317
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329317 2.5 kg 2.5kg
11349317 25 g 25g
11339317 100 g 100g
11319317 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11329317 Supplier Thermo Scientific Alfa Aesar Supplier No. 040184.A4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use With Enamels, Semiconductors, Grinding Wheels | CAS: 1308-06-1 | Co3O4 | 240.80 g/mol

Finds use in enamels, semiconductors, grinding wheels. Used in catalysis, superconductors, ceramics and other fields as an important inorganic material. Also used as catalyst and catalyst carrier, as electrode active material, for glass, porcelain colorants and pigments, as chemical industry oxidant and as a catalyst for organic synthesis, in senior goggles and other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices, enamels, solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Color Black
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 99.7% (Metals basis)
Solubility Information Soluble in acids and alkalis. Insoluble in water.
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
Formula Weight 240.8
Odor Odorless
Density 6.11
Chemical Name or Material Cobalt(II, III) oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II,III) oxide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.