missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11371859
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cobalt(II) hydroxide is used as a drying agent for paints and varnishes. It acts as an aid to dry the lithographic inks and catalyst in the preparation of battery electrode impregnating solutions. It is also involved in the synthesis of other cobalt compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
IUPAC Name cobalt(2+);dihydroxide
SMILES [OH-].[OH-].[Co++]

Specifications

Melting Point 250°C (decomposition)
Color Pink to Purple
Physical Form Powder
Quantity 100 g
Linear Formula Co(OH)2
Merck Index 14,2442
Solubility Information Soluble in acids and ammonia; Very slightly soluble in water; Insoluble in dilute alkalis.
Formula Weight 92.95
Percent Purity 99.9%
Odor Odorless
Sensitivity Air Sensitive
Assay (metals basis)
Chemical Name or Material Cobalt(II) hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'