missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals

£41.10 - £525.00

Chemical Identifiers

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
IUPAC Name cobalt(2+);dihydroxide
SMILES [OH-].[OH-].[Co++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371859
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012524.22
100 g
£41.10
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11361859
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012524.36
500 g
£146.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11351859
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012524.A3
2 kg
£525.00
2kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) hydroxide is used as a drying agent for paints and varnishes. It acts as an aid to dry the lithographic inks and catalyst in the preparation of battery electrode impregnating solutions. It is also involved in the synthesis of other cobalt compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21041-93-0
92.95
ASKVAEGIVYSGNY-UHFFFAOYSA-L
10129900
[OH-].[OH-].[Co++]
CoH2O2
MFCD00016015
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
cobalt(2+);dihydroxide

Specifications

250°C (decomposition)
Pink to Purple
CoH2O2
MFCD00016015
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
ASKVAEGIVYSGNY-UHFFFAOYSA-L
cobalt(2+);dihydroxide
10129900
99.9%
Air Sensitive
Cobalt(II) hydroxide
21041-93-0
Powder
Co(OH)2
14,2442
Soluble in acids and ammonia; Very slightly soluble in water; Insoluble in dilute alkalis.
[OH-].[OH-].[Co++]
92.95
92.95
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302+H332-H317-H319-H334-H350

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 244-166-4

missing translation for 'rtecsNumber' : GG0904500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only