missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hydroxide, 97%

Product Code. 11469537
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11469537

Brand: Thermo Scientific Alfa Aesar B22045.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21041-93-0 | CoH2O2 | 92.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
SMILES [OH-].[OH-].[Co++]

Specifications

Melting Point 250°C (decomposition)
Quantity 250 g
Linear Formula Co(OH)2
Merck Index 14,2442
IUPAC Name cobalt(2+);dihydroxide
Formula Weight 92.95
Percent Purity 97%
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Cobalt(II) hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.