missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hydroxide, 97%

Product Code. 11459537
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459537 50 g 50g
11469537 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459537 Supplier Thermo Scientific Alfa Aesar Supplier No. B22045.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21041-93-0 | CoH2O2 | 92.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-93-0
Molecular Formula CoH2O2
Molecular Weight (g/mol) 92.95
MDL Number MFCD00016015
InChI Key ASKVAEGIVYSGNY-UHFFFAOYSA-L
Synonym cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis
PubChem CID 10129900
SMILES [OH-].[OH-].[Co++]

Specifications

Melting Point 250°C (decomposition)
Quantity 50 g
Linear Formula Co(OH)2
Merck Index 14,2442
IUPAC Name λ²-cobalt(2+) dihydroxide
Formula Weight 92.95
Percent Purity 97%
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Cobalt(II) hydroxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt (II) hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Cobalt hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.