missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) hexafluoro-2,4-pentanedionate hydrate, 97%, Thermo Scientific™

Product Code. 11398718
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398718 1 g 1g
11308728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398718 Supplier Thermo Scientific Alfa Aesar Supplier No. 041594.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206986-92-7
Molecular Formula C10H6CoF12O5
Molecular Weight (g/mol) 493.07
MDL Number MFCD00150649,MFCD00042510
InChI Key POHWVOSIFCILEF-UHFFFAOYSA-N
Synonym bis hexafluoroacetylacetonato cobalt ii hydrate, cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
PubChem CID 16212172
IUPAC Name cobalt;(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
SMILES O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 173°C to 179°C
Color Orange
Physical Form Powder
Quantity 1 g
Solubility Information It is insoluble in water.
Formula Weight 473.04 (Anhydrous)
Percent Purity 97%
Odor Odorless
Chemical Name or Material Cobalt(II) hexafluoro-2,4-pentanedionate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.