missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) fluoride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15436135
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a catalyst to alloy metals and also used for optical deposition, of which it tremendously improves optical quality, in dental care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-17-2
Molecular Formula CoF2
Molecular Weight (g/mol) 96.93
MDL Number MFCD00010941
InChI Key YCYBZKSMUPTWEE-UHFFFAOYSA-L
Synonym cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
PubChem CID 24820
IUPAC Name difluorocobalt
SMILES F[Co]F

Specifications

Boiling Point 1400°C
Melting Point 1200°C
Physical Form Powder
Quantity 50 g
UN Number UN2923
Merck Index 14,2440
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 96.93
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.46 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) fluoride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'